Learn More
Medchemexpress LLC Cyanine5.5 NHS ester (tetrafluoroborate) | 2375105-86-3 | 98.4% | 767.66 | 25 MG
SDP
Catalog No. 5000366179
Encompass_Preferred
Cyanine5.5 NHS ester tetrafluoroborate belongs to the cyanine dye series and is a common fluorescent marker for biomolecules that can interact with biomolecules. Cyanine dyes may also bind to double-helical DNA through intercalation and exhibit enhanced fluorescence upon binding.
  • Belongs to the cyanine dye series
  • Common fluorescent marker for biomolecules
  • Interacts with biomolecules
  • Binds to double-helical DNA through intercalation
  • Exhibits enhanced fluorescence upon binding
  • For research use only
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 50 mg/mL (65.13 mM)
Catalog No. 50-003-66179 Supplier Medchemexpress LLC Supplier No. 25MGHYD1322
Cyanine5.5 NHS ester tetrafluoroborate belongs to the cyanine dye series and is a common fluorescent marker for biomolecules that can interact with biomolecules. Cyanine dyes may also bind to double-helical DNA through intercalation and exhibit enhanced fluorescence upon binding.
  • Belongs to the cyanine dye series
  • Common fluorescent marker for biomolecules
  • Interacts with biomolecules
  • Binds to double-helical DNA through intercalation
  • Exhibits enhanced fluorescence upon binding
  • For research use only
  • Powder can be stored at -20°C for 3 years
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 50 mg/mL (65.13 mM)