Learn More
Medchemexpress LLC Cyanine5 alkyne | 1223357-57-0 | 98.2% | 556.18 g/mol | C35H42ClN3O | 5 MG
SDP
Catalog No. 5000222410
Encompass_Preferred
Cyanine5 alkyne is a fluorescent Cy5 dye bearing an alkyne functional group for bioorthogonal labeling of azide-containing biomolecules via click chemistry. Supplied as a solid typically dissolved in DMSO, it is used for conjugation, imaging, and analysis of protein modifications in biochemical and cell-based assays.
  • Alkyne group enables copper-catalyzed azide-alkyne cycloaddition for selective conjugation.
  • Cy5 spectral properties allow detection in the far-red fluorescence channel.
  • High purity (98.2%) supports reliable labeling performance.
  • Soluble in DMSO for easy preparation of labeling solutions.
  • Suitable for fluorescent imaging and analysis of post-translational modifications.
Catalog No. 50-002-22410 Supplier Medchemexpress LLC Supplier No. HY1370425MG
Cyanine5 alkyne is a fluorescent Cy5 dye bearing an alkyne functional group for bioorthogonal labeling of azide-containing biomolecules via click chemistry. Supplied as a solid typically dissolved in DMSO, it is used for conjugation, imaging, and analysis of protein modifications in biochemical and cell-based assays.
  • Alkyne group enables copper-catalyzed azide-alkyne cycloaddition for selective conjugation.
  • Cy5 spectral properties allow detection in the far-red fluorescence channel.
  • High purity (98.2%) supports reliable labeling performance.
  • Soluble in DMSO for easy preparation of labeling solutions.
  • Suitable for fluorescent imaging and analysis of post-translational modifications.