Learn More
Medchemexpress LLC Cyanine5 alkyne | 1223357-57-0 | 98.2% | C35H42ClN3O | 10MG
SDP
Catalog No. 5000259101
Encompass_Preferred
Cyanine5 alkyne is a near-infrared fluorescent dye bearing a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (click chemistry). It enables covalent labeling of azide-functionalized biomolecules for fluorescence imaging and detection in biochemical assays.
  • Click-reactive terminal alkyne enables CuAAC conjugation.
  • Near-infrared excitation and emission for reduced background in imaging.
  • High purity suitable for sensitive fluorescence assays.
  • Available in multiple milligram sizes for flexible experimental use.
  • Stable when stored sealed away from moisture and light.
Catalog No. 50-002-59101 Supplier Medchemexpress LLC Supplier No. HY13704210MG
Cyanine5 alkyne is a near-infrared fluorescent dye bearing a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (click chemistry). It enables covalent labeling of azide-functionalized biomolecules for fluorescence imaging and detection in biochemical assays.
  • Click-reactive terminal alkyne enables CuAAC conjugation.
  • Near-infrared excitation and emission for reduced background in imaging.
  • High purity suitable for sensitive fluorescence assays.
  • Available in multiple milligram sizes for flexible experimental use.
  • Stable when stored sealed away from moisture and light.