Learn More
Medchemexpress LLC Cyanine5 alkyne (Alkyne-Cy5) | 1223357-57-0 | 98.2% | 556.18 | 50 MG
SDP
Catalog No. 5000349025
Encompass_Preferred
Cyanine5 alkyne (Alkyne-Cy5) is a fluorescent dye utilized for labeling azide proteins, enabling analysis of post-translational modifications like glycosylation. It also functions as a mitochondrial OXPHOS inhibitor, hindering the growth of cancer stem cells (CSCs). This compound is a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • Enables analysis of post-translational modifications like glycosylation
  • Functions as a mitochondrial OXPHOS inhibitor
  • Hinders the growth of cancer stem cells (CSCs)
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Significantly inhibits 3D mammosphere formation in MCF7 cells
Catalog No. 50-003-49025 Supplier Medchemexpress LLC Supplier No. HY13704250MG
Cyanine5 alkyne (Alkyne-Cy5) is a fluorescent dye utilized for labeling azide proteins, enabling analysis of post-translational modifications like glycosylation. It also functions as a mitochondrial OXPHOS inhibitor, hindering the growth of cancer stem cells (CSCs). This compound is a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • Enables analysis of post-translational modifications like glycosylation
  • Functions as a mitochondrial OXPHOS inhibitor
  • Hinders the growth of cancer stem cells (CSCs)
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Significantly inhibits 3D mammosphere formation in MCF7 cells