Learn More
Lumiprobe CYANINE5 AMINE, 25MG
Catalog No. NC0775121
Encompass_Preferred
Red emitting fluorescent dye with a free amino group for the reaction with activated esters, activated carboxy groups, or epoxides
Catalog No. NC0775121 Supplier Lumiprobe Supplier No. 430C0
Red emitting fluorescent dye with a free amino group for the reaction with activated esters, activated carboxy groups, or epoxides