Learn More
Medchemexpress LLC Cyanine5 DBCO hexafluorophosphate | 2360411-64-7 | 98.1% | 929.03 | C53H59F6N4O2P | 10 MG
SDP
Catalog No. 5000297236
Encompass_Preferred
Cyanine5 DBCO hexafluorophosphate is a near-infrared azide-reactive fluorescent probe for copper-free click chemistry (SPAAC). It is used to label and track azide-modified biomolecules and cells in vitro and in vivo, and is supplied as a solid with reported high purity.
Catalog No. 50-002-97236 Supplier Medchemexpress LLC Supplier No. HYD158310MG
Cyanine5 DBCO hexafluorophosphate is a near-infrared azide-reactive fluorescent probe for copper-free click chemistry (SPAAC). It is used to label and track azide-modified biomolecules and cells in vitro and in vivo, and is supplied as a solid with reported high purity.