Learn More
Lumiprobe Cyanine5 maleimide | 1437796-65-0 | MFCD28385511 | 25 mg
Catalog No. NC1283454
Encompass_Preferred
Cyanine5 maleimide | Purity: 95% | Mol Wt: 641.24 | 1437796-65-0 | MFCD28385511 | 25 mg
Catalog No. NC1283454 Supplier Lumiprobe Supplier No. 43080
Cyanine5 maleimide | Purity: 95% | Mol Wt: 641.24 | 1437796-65-0 | MFCD28385511 | 25 mg