Learn More
TARGETMOL CHEMICALS INC Cyanine5 NHS ester chloride
SDP
Catalog No. 502878983
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cyanine5 NHS ester chloride is a reactive dye with red fluorescence that requires a small amount of organic cosolvent (such as DMF or DMSO) to label amino groups in peptides, proteins, and oligonucleotides. Purity 99.06%
Catalog No. 50-287-8983 Supplier TARGETMOL CHEMICALS INC Supplier No. T384325MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cyanine5 NHS ester chloride is a reactive dye with red fluorescence that requires a small amount of organic cosolvent (such as DMF or DMSO) to label amino groups in peptides, proteins, and oligonucleotides. Purity 99.06%