Learn More
Medchemexpress LLC Cyanine5 NHS ester (iodide) | 00-00-0 | 98.0% | C36H42IN3O4 | 10MG
SDP
Catalog No. 5000260182
Encompass_Preferred
Cyanine5 NHS ester iodide is an amine-reactive, red-emitting fluorescent dye for covalent labeling of primary amino groups in peptides, proteins, and oligonucleotides. Supplied as a solid NHS-ester with an iodide counter-ion, it is characterized for research use with COA, SDS, HNMR, and LCMS documentation to support conjugation and analytical workflows.
  • Red emission suitable for near-infrared fluorescence detection.
  • Amine-reactive NHS ester for covalent labeling of primary amines.
  • High purity supporting consistent conjugation performance.
  • Available in multiple small-scale pack sizes for research use.
  • Characterized by COA, HNMR, and LCMS documentation.
Catalog No. 50-002-60182 Supplier Medchemexpress LLC Supplier No. HY135414B10MG
Cyanine5 NHS ester iodide is an amine-reactive, red-emitting fluorescent dye for covalent labeling of primary amino groups in peptides, proteins, and oligonucleotides. Supplied as a solid NHS-ester with an iodide counter-ion, it is characterized for research use with COA, SDS, HNMR, and LCMS documentation to support conjugation and analytical workflows.
  • Red emission suitable for near-infrared fluorescence detection.
  • Amine-reactive NHS ester for covalent labeling of primary amines.
  • High purity supporting consistent conjugation performance.
  • Available in multiple small-scale pack sizes for research use.
  • Characterized by COA, HNMR, and LCMS documentation.