Learn More
Hach Company CyaniVer 3 Cyanide Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501995412 Shop All Hach Company Products
Encompass
For cyanide determination by the Pyridine-Pyrazolone Method. For instruments requiring a 10-mL sample.
Catalog No. 50-199-5412 Supplier Hach Company Supplier No. 2106869
For cyanide determination by the Pyridine-Pyrazolone Method. For instruments requiring a 10-mL sample.