Learn More
Hach Company CyaniVer 4 Cyanide Reagent Powder Pillows, 25mL, pk/100
Supplier:  Hach Company 1404099
Encompass
For cyanide determination by the Pyridine-Pyrazolone method (Hach method 8027). Sample size: 25 mL Pack of 100 Powder Pillows
Catalog No. 50-199-4898