Learn More
Hach Company CyaniVer 5 Cyanide Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501995414 Shop All Hach Company Products
Encompass
For cyanide determination by the Pyridine-Pyrazolone Method. For instruments requiring a 10-mL sample.
Catalog No. 50-199-5414 Supplier Hach Company Supplier No. 2107069
For cyanide determination by the Pyridine-Pyrazolone Method. For instruments requiring a 10-mL sample.