Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc CYANO 10UM 120A 100X4.6MM
SDP

Catalog No. 5090005272
Encompass_Preferred

Cyano 10Um; 120A; 100 X 4.6mm

Catalog No. 50-900-05272 Supplier YMC America, Inc Supplier No. CN12S111046WT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyano 10Um; 120A; 100 X 4.6mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.