Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc CYANO 3UM 120A 10X5MM
SDP

Catalog No. 5090005219
Encompass_Preferred

Cyano 3Um; 120A; 10 X 5mm

Catalog No. 50-900-05219 Supplier YMC America, Inc Supplier No. CN12S030105WFG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyano 3Um; 120A; 10 X 5mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.