Learn More
Medchemexpress LLC α-cyano-4-hydroxycinnamic acid | 28166-41-8 | MFCD00004204 | 99.9% | 189.17 | C10H7NO3 | 1 ML
SDP
Catalog No. 5000241305
Encompass_Preferred
α-Cyano-4-hydroxycinnamic acid is a small-molecule inhibitor of monocarboxylate transporters commonly used in biochemical research and as a matrix in mass spectrometry. It is supplied at high purity and is available as both solid material and as a ready-to-use 10 mM solution in DMSO for laboratory use.
  • High purity (99.9%) suitable for sensitive assays.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid in multiple sizes for flexible use.
  • Effective monocarboxylate transporter inhibitor for in vitro studies.
  • Suitable as a MALDI matrix for peptide analysis.
  • Datasheet, certificate of analysis, and safety data sheet available.
Catalog No. 50-002-41305 Supplier Medchemexpress LLC Supplier No. HY10764110MM 1ML
α-Cyano-4-hydroxycinnamic acid is a small-molecule inhibitor of monocarboxylate transporters commonly used in biochemical research and as a matrix in mass spectrometry. It is supplied at high purity and is available as both solid material and as a ready-to-use 10 mM solution in DMSO for laboratory use.
  • High purity (99.9%) suitable for sensitive assays.
  • Available as a 10 mM solution in DMSO for immediate use.
  • Also available as solid in multiple sizes for flexible use.
  • Effective monocarboxylate transporter inhibitor for in vitro studies.
  • Suitable as a MALDI matrix for peptide analysis.
  • Datasheet, certificate of analysis, and safety data sheet available.