Learn More
Medchemexpress LLC α-cyano-4-hydroxycinnamic acid | 28166-41-8 | MFCD00004204 | 99.9% | 189.17 g·mol⁻¹ | C10H7NO3 | 100 MG
SDP
Catalog No. 5000240530
Encompass_Preferred
α-Cyano-4-hydroxycinnamic acid is a cinnamic acid derivative used as a research reagent and biochemical tool. It acts as a potent, non-competitive inhibitor of monocarboxylate transporters (MCTs) and is commonly used as a matrix in MALDI mass spectrometry and in studies of lactate transport and metabolism.
  • Potent non-competitive inhibitor of monocarboxylate transporters.
  • Suitable as a MALDI matrix for peptide analysis.
  • High purity, 99.9% (reported 99.92%).
  • Molecular weight 189.17 g·mol⁻¹.
  • Supplied as a solid for laboratory use.
Catalog No. 50-002-40530 Supplier Medchemexpress LLC Supplier No. HY107641100MG
α-Cyano-4-hydroxycinnamic acid is a cinnamic acid derivative used as a research reagent and biochemical tool. It acts as a potent, non-competitive inhibitor of monocarboxylate transporters (MCTs) and is commonly used as a matrix in MALDI mass spectrometry and in studies of lactate transport and metabolism.
  • Potent non-competitive inhibitor of monocarboxylate transporters.
  • Suitable as a MALDI matrix for peptide analysis.
  • High purity, 99.9% (reported 99.92%).
  • Molecular weight 189.17 g·mol⁻¹.
  • Supplied as a solid for laboratory use.