Learn More
Medchemexpress LLC α-cyano-4-hydroxycinnamic acid | 28166-41-8 | MFCD00004204 | 99.9% | 189.17 g/mol | C10H7NO3 | 50 MG
SDP
Catalog No. 5000240246
Encompass_Preferred
α-Cyano-4-hydroxycinnamic acid is a small-molecule laboratory reagent used as a MALDI matrix for peptide and small-molecule analysis and as a non-competitive inhibitor of monocarboxylate transporters in biochemical research. It is supplied as a high-purity solid and as a DMSO solution, with supporting datasheet and certificate of analysis.
  • Used as a matrix for MALDI mass spectrometry.
  • Acts as a non-competitive monocarboxylate transporter inhibitor.
  • High purity (99.9%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular formula C10H7NO3 and molecular weight 189.17 g/mol.
  • Accompanied by datasheet and certificate of analysis.
Catalog No. 50-002-40246 Supplier Medchemexpress LLC Supplier No. HY10764150MG
α-Cyano-4-hydroxycinnamic acid is a small-molecule laboratory reagent used as a MALDI matrix for peptide and small-molecule analysis and as a non-competitive inhibitor of monocarboxylate transporters in biochemical research. It is supplied as a high-purity solid and as a DMSO solution, with supporting datasheet and certificate of analysis.
  • Used as a matrix for MALDI mass spectrometry.
  • Acts as a non-competitive monocarboxylate transporter inhibitor.
  • High purity (99.9%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular formula C10H7NO3 and molecular weight 189.17 g/mol.
  • Accompanied by datasheet and certificate of analysis.