Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Cyano-4-hydroxycinnamic acid matrix substance for MALDI-MS, MilliporeSigma™ Supelco™

Catalog No. 111013813 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3813 10 x 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3813 Supplier MilliporeSigma Supelco Supplier No. 3946810X10MGF
Add to Cart
Add to Cart

α-Cyano-4-hydroxycinnamic acid (α-CHCA) functions as matrix and helps in facilitating ionization of proteins and peptides in a matrix-assisted laser desorption/ionization (MALDI) time-of-flight (TOF) mass spectrometer.

TRUSTED_SUSTAINABILITY

Specifications

CAS 28166-41-8
Linear Formula HOC6H4 CH=C(CN)CO2 H
MDL Number MFCD00004204
Melting Point 245°C to 250°C; 245°C to 250°C
Synonym alpha-CCA; alpha-CHCA; alpha-Cyano; 4-HCCA; ACCA
Solubility Soluble in methanol
Type Matrix Substance for MALDI-MS
Quantity 10 x 10 mg
Formula Weight 189.17
Grade Matrix Substance for MALDI-MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.