Learn More
YMC America, Inc CYANO 5UM 120A 50X2MM
SDP
Catalog No. 50157004
Encompass_Preferred
CN12S05-0502WT: Cyano, 5um, 120A, 50X2mm Column
Catalog No. 50-157-004 Supplier YMC America, Inc Supplier No. CN12S050502WT
CN12S05-0502WT: Cyano, 5um, 120A, 50X2mm Column