Learn More
YMC America, Inc CYANO 5UM 120A 50X3MM
SDP
Catalog No. 50157043
Encompass_Preferred
CN12S05-0503WT: Cyano, 5um, 120A, 50X3mm Column
Catalog No. 50-157-043 Supplier YMC America, Inc Supplier No. CN12S050503WT
CN12S05-0503WT: Cyano, 5um, 120A, 50X3mm Column