Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Cyano-βphenylacrylic Acid, MP Biomedicals

Catalog No. ICN213740 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213740 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213740 Supplier MP Biomedicals, Inc Supplier No. 0521374005

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1011-92-3
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
InChI Key CDUQMGQIHYISOP-UHFFFAOYSA-N
Synonym 2-cyano-3-phenylacrylic acid, alpha-cyanocinnamicacid, cyanocinnamic acid, 2-cyano-3-phenyl-2-propenoic acid, acmc-1b4v0, a-cyano-b-phenylacrylic acid, alpha-cyano-beta-phenyl-acrylic acid
PubChem CID 70534
IUPAC Name 2-cyano-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=C(C#N)C(=O)O

Specifications

Quantity 5 g
Chemical Name or Material α-Cyano-β-Phenylacrylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.