Learn More
Analytical Sales & Services Cyano SPE 96-Well Plate, 2.0mL/100mg Bed
Catalog No. 5000806766
Encompass
CN (cyanol) SPE, 2mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-06766 Supplier Analytical Sales & Services Supplier No. 9620CN100
CN (cyanol) SPE, 2mL 96 Well Filter Plate, 100mg per well