Learn More
Analytical Sales & Services Cyano SPE 96-Well Plate, 2.0mL/50mg Bed
Catalog No. 5000806548
Encompass
CN (cyanol) SPE, 2mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-06548 Supplier Analytical Sales & Services Supplier No. 9620CN50
CN (cyanol) SPE, 2mL 96 Well Filter Plate, 50mg per well