Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanoacetic acid, 97%

Catalog No. AAA110450I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA110450I 5000 g
AAA1104530 250 g
AAA110450B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA110450I Supplier Thermo Scientific Chemicals Supplier No. A110450I
Only null left
Add to Cart

CAS: 372-09-8 | C3H3NO2 | 85.062 g/mol

Cyanoacetic acid is used in pharmaceutical industry for producing vitamin B6 and caffeine. It is also used in making dyes, agricultural chemicals, and in the synthesis of phenylacetic acid esters. It acts as a precursor cyanoacrylates viz. ethyl cyanoacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyanoacetic acid is used in pharmaceutical industry for producing vitamin B6 and caffeine. It is also used in making dyes, agricultural chemicals, and in the synthesis of phenylacetic acid esters. It acts as a precursor cyanoacrylates viz. ethyl cyanoacrylate.

Solubility
Miscible with water, ethyl alcohol, ether. Slightly miscible with benzene, acetic acid and chloroform.

Notes
Hygroscopic. Incompatible with strong bases, acids, strong oxidizing agents, alkaline, nitriles and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-09-8
Molecular Formula C3H3NO2
Molecular Weight (g/mol) 85.062
MDL Number MFCD00002677
InChI Key MLIREBYILWEBDM-UHFFFAOYSA-N
Synonym cyanoacetic acid, acetic acid, cyano, malonic mononitrile, acide cyanacetique, cyanessigsaeure, monocyanoacetic acid, malonic acid mononitrile, usaf kf-17, cyanoethanoic acid, kyselina kyanoctova
PubChem CID 9740
ChEBI CHEBI:51889
IUPAC Name 2-cyanoacetic acid
SMILES C(C#N)C(=O)O

Specifications

Melting Point 64°C to 69°C
pH 1.5
Boiling Point 108°C (15 mmHg)
Flash Point 107°C (224°F)
Odor Odorless
Quantity 5000 g
UN Number UN3261
Beilstein 506325
Sensitivity Hygroscopic
Merck Index 14,2689
Solubility Information Miscible with water,ethyl alcohol,ether. Slightly miscible with benzene,acetic acid and chloroform.
Formula Weight 85.06
Percent Purity 99%
Chemical Name or Material Cyanoacetic acid
Show More Show Less
Hazard Category H302+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-743-9
RTECSNumber AG3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.