Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanoacetohydrazide | 140-87-4 | 99.7% | 25 G
SDP

Catalog No. 5000407079
Encompass_Preferred

Cyanoacetohydrazide is a pharmaceutical intermediate. It can be used to synthesize acylhydrazide hydrazone derivatives with antibacterial or anti-tumor activity.

  • Molecular weight of 99.09
  • Chemical formula: C3H5N3O
  • Appears as a solid
  • Color ranges from light yellow to light brown
  • Solid storage at 4°C, sealed, away from moisture
  • Solvent storage at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
  • Soluble in DMSO at 100 mg/mL
  • Soluble in water at 100 mg/mL

Catalog No. 50-004-07079 Supplier Medchemexpress LLC Supplier No. 25GHYB0994
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanoacetohydrazide is a pharmaceutical intermediate. It can be used to synthesize acylhydrazide hydrazone derivatives with antibacterial or anti-tumor activity.

  • Molecular weight of 99.09
  • Chemical formula: C3H5N3O
  • Appears as a solid
  • Color ranges from light yellow to light brown
  • Solid storage at 4°C, sealed, away from moisture
  • Solvent storage at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture
  • Soluble in DMSO at 100 mg/mL
  • Soluble in water at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.