Learn More
Electron Microscopy Sciences EMS EMS Cyanoacrylate Bonder/Debonder Kit, 2x20ml
SDP
Catalog No. 501929865
Encompass_Preferred
The adhesive is a clear, fast curing cyanoacrylate adhesive developed as an alternative for bonding samples and non-porous materials to similar materials or glass. Bonded samples are easy to remove using the remover. The remover is a clear liquid for destabilizing and debonding cured cyanoacrylates. It works on Loctite, Henkel, Permabond, and 3M cyanoacrylate glues. Helpful for cleaning cyanoacylates from surfaces and containers.
Test first before using on plastic. Avoid repeated contact with skin. Includes adhesive and remover, each in 20 ml applicator-tipped bottles.
Catalog No. 50-192-9865 Supplier Electron Microscopy Sciences Supplier No. 12647
The adhesive is a clear, fast curing cyanoacrylate adhesive developed as an alternative for bonding samples and non-porous materials to similar materials or glass. Bonded samples are easy to remove using the remover. The remover is a clear liquid for destabilizing and debonding cured cyanoacrylates. It works on Loctite, Henkel, Permabond, and 3M cyanoacrylate glues. Helpful for cleaning cyanoacylates from surfaces and containers.
Test first before using on plastic. Avoid repeated contact with skin. Includes adhesive and remover, each in 20 ml applicator-tipped bottles.