Learn More
Air Science Usa Cyanoacrylate Fuming Chamber, CA48S, 48" / 1200mm Wide, 115V 60Hz
SDP
Supplier:  Air Science Usa CA48S
Encompass_Preferred
Safefume Cyanoacrylate Fuming Chamber CA48S, 48" Wide. Safefume cyanoacrylate fuming chambers are designed to safely develop latent fingerprints using ethyl cyanoacrylate (cna) vapor in a controlled environment for optimum effectiveness and safety where moisture and fuming time are critical factors. The tamper resistant compartment helps maintain the chain of custody. The unique Multiplex filtration system, together with professional design and unique construction features offer personnel protection during use. The ductless filtration system requires no connection to an outside exhaust system. The automatic control system programs the fuming cycle. A versatile system of hanging rods and shelves allows proper positioning of items of evidence in the chamber. The professionally designed automatic fuming chamber eliminates reliance on do-it-yourself systems that lack user-safety allowances and microprocessor controls needed for repeatable results
Catalog No. NC1537959
Add to Cart
Request a Quote