Learn More
Bioworld CyanoBlue Universal Dye, 5 mL
Catalog No. 50196775
Encompass_Preferred
Premixed and ready-to-load gel dyes. Glow Loading DyesT generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. CyanoBlue dye contains bromophenol blue and xylene cyanol in a ficoll-based solution (6X concentration).
Catalog No. 50-196-775 Supplier Bioworld Supplier No. 105700061
Premixed and ready-to-load gel dyes. Glow Loading DyesT generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. CyanoBlue dye contains bromophenol blue and xylene cyanol in a ficoll-based solution (6X concentration).