Learn More
Sigma Aldrich Fine Chemicals Biosciences Cyanocobalamin Pharmaceutical Secondary Standard; Certified Reference Material | 68-19-9 | 1G
Catalog No. NC1421051
Encompass
Cyanocobalamin Pharmaceutical Secondary Standard; Certified Reference Material | Mol Wt: 1355.37 | 68-19-9 | 1G
Catalog No. NC1421051 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHR12341G
Cyanocobalamin Pharmaceutical Secondary Standard; Certified Reference Material | Mol Wt: 1355.37 | 68-19-9 | 1G