Learn More
Medchemexpress LLC Cyanocobalamin | 68-19-9 | 99.95% | 50 MG
SDP
Catalog No. 5000406544
Encompass_Preferred
Vitamin B12 (Standard) is an analytical standard of Vitamin B12. It is intended for research and analytical applications, playing a key role in the normal functioning of the brain and nervous system, and for the formation of blood. This compound is of analytical standard grade and serves as a reference standard for assays.
  • Intended for research and analytical applications
  • Plays a key role in brain and nervous system function
  • Important for blood formation
  • Serves as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-06544 Supplier Medchemexpress LLC Supplier No. HYB0315R50MG
Vitamin B12 (Standard) is an analytical standard of Vitamin B12. It is intended for research and analytical applications, playing a key role in the normal functioning of the brain and nervous system, and for the formation of blood. This compound is of analytical standard grade and serves as a reference standard for assays.
  • Intended for research and analytical applications
  • Plays a key role in brain and nervous system function
  • Important for blood formation
  • Serves as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS