Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanocobalamin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015155 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5155 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5155 Supplier MilliporeSigma Supelco Supplier No. PHR12341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Cyanocobalamin, also known as vitamin B12, belongs to a group of organic compounds important for the normal growth and development of human and animal bodies. It is used for the treatment of trigeminal neuralgia and multiple sclerosis. Featured Industries: Pharmaceutical (small molecule), Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

CAS 68-19-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C63H88 CoN14O14 P
Synonym Vitamin B12; Cyanocobalmin; Vitamin B12; alpha-(5,6-Dimethylbenzimidazolyl)cyanocobamide; CN-Cbl; Cyanocob(III)alamin; Cyanocobalamin
Type Certified Reference Material
Quantity 1 g
Formula Weight 1355.37
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.