Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Cyanocobalamin, USP (Vitamin B12) (Dietary Supplement Grade)
SDP

Catalog No. 14185552 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-552 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-552 Supplier Medisca Supplier No. 046803
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanocobalamin, USP (Vitamin B12) (Dietary Supplement Grade) ranges from dark red crystals to a amorphous or crystalline red powder. In the anhydrous form, it is very hygroscopic.

Cyanocobalamin, USP (Vitamin B12) ranges from dark red crystals to a amorphous or crystalline red powder. In the anhydrous form, it is very hygroscopic.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Cobinamide, Vitamin B12

Specifications

Chemical Name or Material Cyanocobalamin, USP (Vitamin B12) (Dietary Supplement Grade)
Packaging Bottle, 15mL, Amber Glass Wide Mouth Packer, 28-400
CAS 68-19-9
Type Drug Standard
Molecular Weight (g/mol) 1359.4108
Quantity 5 g
Percent Purity 96 to 102.0 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.