Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen bromide, 3M solution in dichloromethane, AcroSeal™

Catalog No. AC428911000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC428911000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428911000 Supplier Thermo Scientific Chemicals Supplier No. 428911000
  • $138.00
    $111.65 / Each of 1

    Save $26.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 506-68-3 | BrCN | 105.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyanogen bromide
Color Colorless to Yellow
CAS 506-68-3, 75-09-2
Physical Form Liquid
Quantity 100 mL
Packaging AcroSeal glass Bottle packed in tin can
Specific Gravity 1.44
Density 1.4400g/mL
Molecular Formula BrCN
MDL Number MFCD00011597
Merck Index 15, 2683
Synonym cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn
InChI Key ATDGTVJJHBUTRL-UHFFFAOYSA-N
SMILES C(#N)Br
IUPAC Name carbononitridic bromide
Molecular Weight (g/mol) 105.93
PubChem CID 10476
Formula Weight 105.93
Concentration or Composition (by Analyte or Components) 2.95 to 3.30M
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.