Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanogen Bromide, MP Biomedicals

Catalog No. ICN19031425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19031425 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19031425 Supplier MP Biomedicals, Inc Supplier No. 0219031425

Reagent for selective peptide cleavage

TRUSTED_SUSTAINABILITY

Specifications

CAS 506-68-3
Molecular Formula CBrN
Quantity 25 g
Synonym cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn
InChI Key ATDGTVJJHBUTRL-UHFFFAOYSA-N
SMILES C(#N)Br
IUPAC Name carbononitridic bromide
Molecular Weight (g/mol) 105.922
PubChem CID 10476
Formula Weight 105.9
Chemical Name or Material Cyanogen Bromide
Show More Show Less
storageNote1 Available in U.S.A. only; Not for export.
Recommended Storage 0°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.