Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Cyanomethyl)triphenylphosphonium chloride, 98+%

Catalog No. AAA1309622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1309622 100 g
AAA1309614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1309622 Supplier Thermo Scientific Chemicals Supplier No. A1309622
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4336-70-3 | C20H17ClNP | 337.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4336-70-3
Molecular Formula C20H17ClNP
Molecular Weight (g/mol) 337.79
MDL Number MFCD00031672
InChI Key ARPLQAMUUDIHIT-UHFFFAOYSA-M
Synonym cyanomethyl triphenylphosphonium chloride, phosphonium, cyanomethyl triphenyl-, chloride, cyanomethyl triphenylphosphanium chloride, cyanomethyl triphenyl phosphanium chloride, cyanomethyltriphenylphosphonium chloride, acmc-1ctgb, wln: nc1pr&r&r &g, cyanomethyl triphenylphosphoniumchloride, cyanomethyl triphenyl phosphonium chloride
PubChem CID 197040
IUPAC Name cyanomethyl(triphenyl)phosphanium;chloride
SMILES [Cl-].N#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼270°C (decomposition)
Quantity 100 g
UN Number UN3439
Beilstein 6222047
Sensitivity Hygroscopic
Formula Weight 337.79
Percent Purity ≥98%
Chemical Name or Material (Cyanomethyl)triphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 224-383-0
RTECSNumber TA2075000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.