Learn More
AdipoGen Cyanophos
SDP
Supplier:  AdipoGen CDXC0604M025
Encompass_Preferred
Chemical. CAS 2636-26-2. Formula C9H10NO3PS. MW 243.22. Cyanophos is an organophosphate insecticide, which is effectively used against various root-parasitic nematodes such as Diaspididae, Lepidoptera, Coccidae, Aphididae and other insects on various fruits and vegetables. It is a acetylcholinesterase AChE inhibitor and antagonist of NMDA receptors. It shows a moderate to high level of toxicity to fish, algae and daphnia. Compound can be used as analytical reference material.
Catalog No. 50-256-1356