Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanosafracin B | 96996-50-8 | 96.0% | 549.62 g·mol⁻¹ | C29H35N5O6 | 5 MG
SDP

Catalog No. 5000221981
Encompass_Preferred

Cyanosafracin B is a small-molecule intermediate used in the synthesis of antitumor agents such as ecteinascidin ET-743 and phthalascidin Pt-650. It is supplied as a solid reagent intended for medicinal chemistry and synthetic organic applications.

  • Used as a starting material for synthesis of complex antitumor agents
  • Solid form for convenient handling and storage
  • Molecular weight 549.62 g·mol⁻¹
  • Purity listed as 96.0%
  • Available in small research quantities for early-stage studies

Catalog No. 50-002-21981 Supplier Medchemexpress LLC Supplier No. HY1377875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanosafracin B is a small-molecule intermediate used in the synthesis of antitumor agents such as ecteinascidin ET-743 and phthalascidin Pt-650. It is supplied as a solid reagent intended for medicinal chemistry and synthetic organic applications.

  • Used as a starting material for synthesis of complex antitumor agents
  • Solid form for convenient handling and storage
  • Molecular weight 549.62 g·mol⁻¹
  • Purity listed as 96.0%
  • Available in small research quantities for early-stage studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.