Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyanosafracin B | 96996-50-8 | 96.0% | C29H35N5O6 | 10MG
SDP

Catalog No. 5000259078
Encompass_Preferred

Cyanosafracin B is a small-molecule intermediate used in research and medicinal chemistry as a starting material for the synthesis of antitumor agents such as Ecteinascidin ET-743 and Phthalascidin Pt-650. It is supplied as a solid with defined purity and storage recommendations to maintain stability for laboratory use.

  • Starting material for synthesis of antitumor agents.
  • Chemical formula C29H35N5O6; molecular weight 549.62.
  • Typical purity around 96.0% as supplied for research use.
  • Solid form, light yellow to yellow, suitable for handling in lab settings.
  • Recommended storage: solid at 4°C, protect from light; in solution store frozen.
  • Available in small pack sizes convenient for medicinal chemistry workflows.

Catalog No. 50-002-59078 Supplier Medchemexpress LLC Supplier No. HY13778710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanosafracin B is a small-molecule intermediate used in research and medicinal chemistry as a starting material for the synthesis of antitumor agents such as Ecteinascidin ET-743 and Phthalascidin Pt-650. It is supplied as a solid with defined purity and storage recommendations to maintain stability for laboratory use.

  • Starting material for synthesis of antitumor agents.
  • Chemical formula C29H35N5O6; molecular weight 549.62.
  • Typical purity around 96.0% as supplied for research use.
  • Solid form, light yellow to yellow, suitable for handling in lab settings.
  • Recommended storage: solid at 4°C, protect from light; in solution store frozen.
  • Available in small pack sizes convenient for medicinal chemistry workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.