Learn More
Medchemexpress LLC Cyanox CY 1790 100g | 40601-76-1 | 100 G
SDP
Catalog No. 5000226185
Encompass_Preferred
Cyanox 1790 is a triazine-based hindered phenolic antioxidant used to provide thermal and oxidative stabilization for polyolefins, particularly polyethylene. It offers resistance to chlorine dioxide, low color contribution, and long-term processing stability, making it suitable for research and industrial applications focused on polymer anti-degradation.
  • Provides effective free radical scavenging for long-term thermal stabilization.
  • Improves resistance to chlorine dioxide and gas-fade during service.
  • Contributes low color to polymer formulations, minimizing discoloration.
  • Exhibits good resistance to extraction and migration, enhancing durability.
  • Typical physical form: cream to off-white free-flowing powder for easy handling.
  • Typical purity approximately 99.4% as supplied by common distributors.
Catalog No. 50-002-26185 Supplier Medchemexpress LLC Supplier No. HY116772100G
Add to Cart
Add to Cart
Cyanox 1790 is a triazine-based hindered phenolic antioxidant used to provide thermal and oxidative stabilization for polyolefins, particularly polyethylene. It offers resistance to chlorine dioxide, low color contribution, and long-term processing stability, making it suitable for research and industrial applications focused on polymer anti-degradation.
  • Provides effective free radical scavenging for long-term thermal stabilization.
  • Improves resistance to chlorine dioxide and gas-fade during service.
  • Contributes low color to polymer formulations, minimizing discoloration.
  • Exhibits good resistance to extraction and migration, enhancing durability.
  • Typical physical form: cream to off-white free-flowing powder for easy handling.
  • Typical purity approximately 99.4% as supplied by common distributors.