Learn More
Medchemexpress LLC Cyanox CY 1790 10mM 1mL | 40601-76-1 | 699.9 g/mol | C42H57N3O6 | 1 ML
SDP
Catalog No. 5000228898
Encompass_Preferred
Cyanox CY 1790 is a triazine-based, sterically hindered phenolic antioxidant used to improve oxidative and thermal stability of polyolefins and other engineering plastics. It is supplied as a 10 mM solution in a 1 mL vial for laboratory research and material degradation studies.
  • Triazine-based hindered phenolic antioxidant for polymer stabilization
  • Provides processing and thermal stability during extrusion and molding
  • Offers resistance to gas-fade (chlorine dioxide) and reduced extraction
  • Low color contribution for color-sensitive applications
  • Applicable to polyethylene, polypropylene, polyurethane, and polyester
  • Supplied as a 10 mM solution in a 1 mL vial for research use
Catalog No. 50-002-28898 Supplier Medchemexpress LLC Supplier No. HY11677210MM 1ML
Cyanox CY 1790 is a triazine-based, sterically hindered phenolic antioxidant used to improve oxidative and thermal stability of polyolefins and other engineering plastics. It is supplied as a 10 mM solution in a 1 mL vial for laboratory research and material degradation studies.
  • Triazine-based hindered phenolic antioxidant for polymer stabilization
  • Provides processing and thermal stability during extrusion and molding
  • Offers resistance to gas-fade (chlorine dioxide) and reduced extraction
  • Low color contribution for color-sensitive applications
  • Applicable to polyethylene, polypropylene, polyurethane, and polyester
  • Supplied as a 10 mM solution in a 1 mL vial for research use