Learn More
Medchemexpress LLC Cyanox CY 1790 25g | 40601-76-1 | 699.9 g/mol | C42H57N3O6 | 25 G
SDP
Catalog No. 5000226155
Encompass_Preferred
Cyanox CY 1790 is a hindered phenolic antioxidant (CAS 40601-76-1) used to protect polyolefins and other polymers from oxidative and chlorine-dioxide-type degradation. It is supplied as a white to off-white solid and is intended for research and additive formulation studies.
  • Provides antioxidant protection for polyethylene and other polyolefins
  • Offers resistance to chlorine dioxide-type degradation
  • Supplied as a white to off-white solid suitable for lab use
  • Reported high purity for analytical and research applications (~99.4%)
  • Molecular weight 699.9 g/mol, formula C42H57N3O6
  • Commonly used in studies of polymer stabilization and anti-degradation
Catalog No. 50-002-26155 Supplier Medchemexpress LLC Supplier No. HY11677225G
Cyanox CY 1790 is a hindered phenolic antioxidant (CAS 40601-76-1) used to protect polyolefins and other polymers from oxidative and chlorine-dioxide-type degradation. It is supplied as a white to off-white solid and is intended for research and additive formulation studies.
  • Provides antioxidant protection for polyethylene and other polyolefins
  • Offers resistance to chlorine dioxide-type degradation
  • Supplied as a white to off-white solid suitable for lab use
  • Reported high purity for analytical and research applications (~99.4%)
  • Molecular weight 699.9 g/mol, formula C42H57N3O6
  • Commonly used in studies of polymer stabilization and anti-degradation