Learn More
CTL Scientific Supply Corp Cyantesmo Test Paper, roll of 5 meter length
SDP
Catalog No. 50300316 Shop All CTL Scientific Supply Products
Encompass_Preferred
CYANTESMO PAPER This test paper allows the quick and easy detection of hydrocyanic acid and cyanides in aqueous solutions and extracts. Limit of sensitivity 0.2 mg/l hydrocyanic acid (HCN) 5 meter roll
Catalog No. 50-300-316 Supplier CTL Scientific Supply Corp Supplier No. 90604
CYANTESMO PAPER This test paper allows the quick and easy detection of hydrocyanic acid and cyanides in aqueous solutions and extracts. Limit of sensitivity 0.2 mg/l hydrocyanic acid (HCN) 5 meter roll