Learn More
Medchemexpress LLC Cyantraniliprole-d3 | 1392493-34-3 | 99.6% | 1 MG
SDP
Catalog No. 5000339984
Encompass_Preferred
Cyantraniliprole-d3 is the deuterium labeled Cyantraniliprole, which is an insecticide of the ryanoid class. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-39984 Supplier Medchemexpress LLC Supplier No. HY12779S1MG
Cyantraniliprole-d3 is the deuterium labeled Cyantraniliprole, which is an insecticide of the ryanoid class. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.