Learn More
Hach Company Cyanuric Acid 2 Reagent Powder Pillows, pk/50
Supplier:  Hach Company 246066
Encompass
For determination of cyanuric acid by the melamine turbidimetric method. Hach method 8139. Sample size: 25 mL. Also used in Model CY-3 test kits. Pack of 50 powder pillows.
Catalog No. 50-199-4778