Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanuric acid, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012437 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2437 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2437 Supplier MilliporeSigma Supelco Supplier No. 16614250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyanuric acid (CYA) is a triazine industrial chemical, used in production of scouring powders, household bleaches, industrial cleansers, and automatic dishwasher compounds. It is also used as stabilizers in swimming pools which helps in preventing destruction of chlorine due to UV. It has a weak acid taste and is sparingly soluble in cold water. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-80-5
Physical Form Neat
Linear Formula C3H3N3O3
MDL Number MFCD00082990
Melting Point >360°C
Synonym 1,3,5-Triazine-2,4,6-triol; 2,4,6-Trihydroxy-1,3,5-triazine
Type Analytical Standard
Quantity 250 mg
Formula Weight 129.07
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.