Cyanuric Acid, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar A1544730

Quantity 250g

 View more versions of this product

Catalog No. AAA1544730

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range 99%
    Molecular Formula C3H3N3O3
    Formula Weight 129.08
    Chemical Name or Material Cyanuric Acid

    Specifications

    Formula Weight 129.08
    Quantity 250g
    TSCA TSCA
    MDL Number MFCD00082990
    Beilstein 126982
    Merck Index 14,2698
    RTECS Number XZ1800000
    Assay Percent Range 99%
    Molecular Formula C3H3N3O3
    CAS 108-80-5
    Storage Note 1 Hygroscopic
    Chemical Name or Material Cyanuric Acid
    Assay 99%

    Description

    Solubility
    Soluble in sulfuric acid, dimethylformamide, sodium hydroxide, potassium hydroxide and diemthyl sulfoxide. Slightly soluble in acetone, benzene, diethyl ether, ethanol and hexane. Insoluble in cold methanol, benzene and chloroform.

    Notes
    Store in a cool place. Incompatible with strong oxidizing agents.

    Hygroscopic

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit