Learn More
Aviva Systems Biology Corp CYB561 Antibody - middle region (ARP41992_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41992_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYB561. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYB561. Alternate names: FRRS2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58072