Learn More
Aviva Systems Biology Corp Cyb5r1 Antibody - N-terminal region (ARP47235_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47235_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cyb5r1. It was validated on Western Blot. . Alternate names: 1500005G05Rik, B5R.1, C80155, Nqo3a2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-292