Learn More
Aviva Systems Biology Corp Cyb5r4 Antibody - N-terminal region (ARP56860_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56860_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cyb5r4. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2810034J18Rik, B5+B5R, C79736, Cb5cb5r, Ncb5or, b5/b5r, b5b5rApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-225