Learn More
Aviva Systems Biology Corp CYBA Antibody - middle region (ARP44214_P050) | 100uL
SDP
Catalog No. 5001458747
Encompass_Preferred
This is a rabbit polyclonal antibody against CYBA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYBA. Alternate names: p22-PHOXApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58747 Supplier Aviva Systems Biology Corp Supplier No. ARP44214_P050
This is a rabbit polyclonal antibody against CYBA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CYBA. Alternate names: p22-PHOXApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep